Learn More
Medchemexpress LLC 2,5,8,11,14,17,20,23,26,29,32,35,38,41,44-Pentadecaoxahexatetracontan-46-amine | 2745717-94-4 | 99.9% | 691.85 | 50 MG
SDP
Catalog No. 5000350299
Encompass_Preferred
The m-PEG15-amine is a chemical compound with the molecular formula C31H65NO15 and a molecular weight of 691.85. It is provided as a colorless to light yellow oil with a purity of 99.91%. This product is intended for use as a laboratory chemical and in the manufacture of substances. It is classified as hazardous, causing acute oral toxicity, skin and serious eye irritation, and potential respiratory tract irritation.
  • Consistent with structure based on ¹H NMR Spectrum and LCMS
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation
Catalog No. 50-003-50299 Supplier Medchemexpress LLC Supplier No. HY13842450MG
The m-PEG15-amine is a chemical compound with the molecular formula C31H65NO15 and a molecular weight of 691.85. It is provided as a colorless to light yellow oil with a purity of 99.91%. This product is intended for use as a laboratory chemical and in the manufacture of substances. It is classified as hazardous, causing acute oral toxicity, skin and serious eye irritation, and potential respiratory tract irritation.
  • Consistent with structure based on ¹H NMR Spectrum and LCMS
  • Harmful if swallowed
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation