Learn More
Medchemexpress LLC 2,5,8,11,14,17,20,23,26-Nonaoxaoctacosan-28-amine | 211859-73-3 | 97.0% | 1 G
SDP
Catalog No. 5000339954
Encompass_Preferred
m-PEG9-Amine is a versatile PEG-based linker. It functions as a PROTAC linker, facilitating the synthesis of Proteolysis-Targeting Chimeras (PROTACs). Additionally, it serves as a cleavable ADC linker, crucial for the synthesis of antibody-drug conjugates (ADCs).
  • Functions as a PEG-based PROTAC linker
  • Enables the synthesis of Proteolysis-Targeting Chimeras (PROTACs)
  • Acts as a cleavable ADC linker
  • Essential for the synthesis of antibody-drug conjugates (ADCs)
  • Utilizes the intracellular ubiquitin-proteasome system for targeted protein degradation
Catalog No. 50-003-39954 Supplier Medchemexpress LLC Supplier No. HY1305711G
m-PEG9-Amine is a versatile PEG-based linker. It functions as a PROTAC linker, facilitating the synthesis of Proteolysis-Targeting Chimeras (PROTACs). Additionally, it serves as a cleavable ADC linker, crucial for the synthesis of antibody-drug conjugates (ADCs).
  • Functions as a PEG-based PROTAC linker
  • Enables the synthesis of Proteolysis-Targeting Chimeras (PROTACs)
  • Acts as a cleavable ADC linker
  • Essential for the synthesis of antibody-drug conjugates (ADCs)
  • Utilizes the intracellular ubiquitin-proteasome system for targeted protein degradation