Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,5,8,11,14,17,20,23,26-nonaoxaoctacosan-28-amine | 211859-73-3 | MFCD16619283 | 97.0% | 427.53 | C19H41NO9 | 250 MG
SDP

Catalog No. 5000202508
Encompass_Preferred

m-PEG9-Amine is a methoxy-terminated polyethylene glycol linker bearing a terminal primary amine used in synthesis workflows for antibody-drug conjugates and PROTAC molecules.

  • Used as a cleavable ADC linker and as a PEG-based PROTAC linker.
  • Molecular formula C19H41NO9 and molecular weight 427.53.
  • Chemical name: 2,5,8,11,14,17,20,23,26-nonaoxaoctacosan-28-amine.
  • Purity typically 97.0% by NMR (per COA).
  • Store dissolved in solvent at -80°C (6 months) or -20°C (1 month); protect from light.

Catalog No. 50-002-02508 Supplier Medchemexpress LLC Supplier No. HY130571250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG9-Amine is a methoxy-terminated polyethylene glycol linker bearing a terminal primary amine used in synthesis workflows for antibody-drug conjugates and PROTAC molecules.

  • Used as a cleavable ADC linker and as a PEG-based PROTAC linker.
  • Molecular formula C19H41NO9 and molecular weight 427.53.
  • Chemical name: 2,5,8,11,14,17,20,23,26-nonaoxaoctacosan-28-amine.
  • Purity typically 97.0% by NMR (per COA).
  • Store dissolved in solvent at -80°C (6 months) or -20°C (1 month); protect from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.