Learn More
Medchemexpress LLC (2,5,8,11,14,17,20,23,26-nonaoxaoctacosan-28-yl)phosphonic acid | 2055016-25-4 | MFCD28505502 | 99.3% | C19H41O12P | 10MG
SDP
Catalog No. 5000259301
Encompass_Preferred
m-PEG9-phosphonic acid is a methoxy-terminated polyethylene glycol (PEG) linker bearing a terminal phosphonic acid. It is intended for use as a spacer and conjugation handle in the synthesis of PROTACs and other bioconjugates for research applications.
  • PEG-based spacer with nine ethylene glycol units.
  • Terminal phosphonic acid functional group for conjugation chemistry.
  • High purity: 99.3%.
  • Molecular weight 492.50 g/mol and formula C19H41O12P.
  • Available in milligram quantities suitable for research-scale synthesis.
Catalog No. 50-002-59301 Supplier Medchemexpress LLC Supplier No. HY13055410MG
m-PEG9-phosphonic acid is a methoxy-terminated polyethylene glycol (PEG) linker bearing a terminal phosphonic acid. It is intended for use as a spacer and conjugation handle in the synthesis of PROTACs and other bioconjugates for research applications.
  • PEG-based spacer with nine ethylene glycol units.
  • Terminal phosphonic acid functional group for conjugation chemistry.
  • High purity: 99.3%.
  • Molecular weight 492.50 g/mol and formula C19H41O12P.
  • Available in milligram quantities suitable for research-scale synthesis.