Learn More
Medchemexpress LLC 2,5,8,11,14-pentaoxahexadecan-16-ol, 1-phenyl | 57671-28-0 | MFCD06797154 | 98.6% | 328.40 g/mol | C17H28O6 | 5 G
SDP
Catalog No. 5000210591
Encompass_Preferred
BnO-PEG5-OH is a benzyl-protected PEG5 primary alcohol used as a short spacer or linker building block in PROTAC and conjugation chemistries. It provides a PEG chain with a terminal hydroxyl group masked by a benzyl protecting group, allowing selective deprotection and downstream functionalization.
  • Provides a short polyethylene glycol spacer for linker design
  • Contains a benzyl-protected terminal hydroxyl for selective deprotection
  • Suitable for building block use in PROTAC and bioconjugate synthesis
  • Available in multiple pack sizes for small-scale synthesis
  • Offers high reported purity for synthetic applications
Catalog No. 50-002-10591 Supplier Medchemexpress LLC Supplier No. HYW0427145G
BnO-PEG5-OH is a benzyl-protected PEG5 primary alcohol used as a short spacer or linker building block in PROTAC and conjugation chemistries. It provides a PEG chain with a terminal hydroxyl group masked by a benzyl protecting group, allowing selective deprotection and downstream functionalization.
  • Provides a short polyethylene glycol spacer for linker design
  • Contains a benzyl-protected terminal hydroxyl for selective deprotection
  • Suitable for building block use in PROTAC and bioconjugate synthesis
  • Available in multiple pack sizes for small-scale synthesis
  • Offers high reported purity for synthetic applications