Learn More
Medchemexpress LLC 2,5,8,11-Tetraoxatetradec-13-yne | 89635-82-5 | 98.0% | 202.25 | 5 G
SDP
Catalog No. 5000351376
Encompass_Preferred
Propargyl-PEG3-methane is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is a click chemistry reagent containing an Alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-003-51376 Supplier Medchemexpress LLC Supplier No. HY1400595G
Propargyl-PEG3-methane is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is a click chemistry reagent containing an Alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups