Learn More
Medchemexpress LLC 2,5,8,11-Tetraoxatridecane, 13-azido- | 606130-90-9 | 98.0% | C9H19N3O4 | 100 MG
SDP
Catalog No. 5000353383
Encompass_Preferred
m-PEG4-azide is a PEG-based PROTAC linker, presented as a colorless to light yellow liquid. This compound has a purity of 98.0%.
  • Consistent with structure as verified by 1H NMR and MS analysis.
  • Store in pure form at -20°C for up to 3 years.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month.
  • Complies with specified analytical data.
Catalog No. 50-003-53383 Supplier Medchemexpress LLC Supplier No. HY140828100MG
m-PEG4-azide is a PEG-based PROTAC linker, presented as a colorless to light yellow liquid. This compound has a purity of 98.0%.
  • Consistent with structure as verified by 1H NMR and MS analysis.
  • Store in pure form at -20°C for up to 3 years.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month.
  • Complies with specified analytical data.