Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,5,8,11-Tetraoxatridecane, 13-azido- | 606130-90-9 | 98.0% | C9H19N3O4 | 50 MG
SDP

Catalog No. 5000355379
Encompass_Preferred

2,5,8,11-Tetraoxatridecane, 13-azido- is a chemical compound with a molecular weight of 233.26. It appears as a colorless to light yellow liquid and is intended for research use only. This product has been tested and complies with the given specifications.

  • Colorless to light yellow liquid appearance
  • Molecular weight of 233.26
  • Suitable for research applications
  • Storage in pure form at -20°C for 3 years
  • Storage in solvent at -80°C for 6 months
  • Storage in solvent at -20°C for 1 month

Catalog No. 50-003-55379 Supplier Medchemexpress LLC Supplier No. HY14082850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,5,8,11-Tetraoxatridecane, 13-azido- is a chemical compound with a molecular weight of 233.26. It appears as a colorless to light yellow liquid and is intended for research use only. This product has been tested and complies with the given specifications.

  • Colorless to light yellow liquid appearance
  • Molecular weight of 233.26
  • Suitable for research applications
  • Storage in pure form at -20°C for 3 years
  • Storage in solvent at -80°C for 6 months
  • Storage in solvent at -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.