Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,5-Anhydro-D-mannitol | 41107-82-8 | MFCD00012443 | 99.9% | 164.16 | C6H12O5 | 10 MG
SDP

Catalog No. 5000300392
Encompass_Preferred

2,5-Anhydro-D-mannitol is a biochemical assay reagent labeled at 97% and verified at 99.86% purity by HPLC. It is a white to off-white solid (C6H12O5, 164.16 g/mol) intended for research use in analytical and biochemical workflows.

  • High purity (99.86% by HPLC).
  • White to off-white solid suitable for routine handling.
  • Molecular formula C6H12O5; molecular weight 164.16 g/mol.
  • Storage: store under nitrogen at 4°C; in solvent: -80°C (6 months), -20°C (1 month).
  • Intended for research use only.

Catalog No. 50-003-00392 Supplier Medchemexpress LLC Supplier No. HYW03931510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,5-Anhydro-D-mannitol is a biochemical assay reagent labeled at 97% and verified at 99.86% purity by HPLC. It is a white to off-white solid (C6H12O5, 164.16 g/mol) intended for research use in analytical and biochemical workflows.

  • High purity (99.86% by HPLC).
  • White to off-white solid suitable for routine handling.
  • Molecular formula C6H12O5; molecular weight 164.16 g/mol.
  • Storage: store under nitrogen at 4°C; in solvent: -80°C (6 months), -20°C (1 month).
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.