Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-[[5-(benzylamino)-1,3,4-thiadiazol-2-yl]sulfanyl]-N-(cyclohexylmethyl)acetamide | 556803-08-8 | 99.2% | 376.54 | C18H24N4OS2 | 50 MG
SDP

Catalog No. 5000240162
Encompass_Preferred

TCN 213 is a selective, surmountable, glycine-dependent GluN1/GluN2A NMDA receptor antagonist used as a research tool to probe NMDA receptor pharmacology and monitor developmental switches in cortical neurons. Supplied as a solid for in vitro pharmacology and biochemical assays.

  • Purity 99.16%.
  • Formula C18H24N4OS2.
  • Molecular weight 376.54 g/mol.
  • CAS number 556803-08-8.
  • Mode of action selective glycine-dependent GluN1/GluN2A NMDA receptor antagonist.
  • Intended use in vitro pharmacology and receptor characterization.

Catalog No. 50-002-40162 Supplier Medchemexpress LLC Supplier No. HY10771250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TCN 213 is a selective, surmountable, glycine-dependent GluN1/GluN2A NMDA receptor antagonist used as a research tool to probe NMDA receptor pharmacology and monitor developmental switches in cortical neurons. Supplied as a solid for in vitro pharmacology and biochemical assays.

  • Purity 99.16%.
  • Formula C18H24N4OS2.
  • Molecular weight 376.54 g/mol.
  • CAS number 556803-08-8.
  • Mode of action selective glycine-dependent GluN1/GluN2A NMDA receptor antagonist.
  • Intended use in vitro pharmacology and receptor characterization.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.