Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Bis(2,2,2-trifluoroethoxy)benzoic acid, 96%, Thermo Scientific™

Catalog No. AAH6110314
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6110314 25 g
AAH6110303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6110314 Supplier Thermo Scientific Chemicals Supplier No. H6110314

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35480-52-5
Molecular Formula C11H8F6O4
Molecular Weight (g/mol) 318.171
MDL Number MFCD00221424
InChI Key YPGYLCZBZKRYQJ-UHFFFAOYSA-N
Synonym 2,5-bis 2,2,2-trifluoroethoxy benzoic acid, 2,5-bis trifluoroethoxy benzoic acid, 2,5-di 2,2,2-trifluoroethoxy benzoic acid, benzoic acid, 2,5-bis 2,2,2-trifluoroethoxy, flecainide impurity d, pubchem10082, acmc-209ih5, 2,5-di 2,2,2-trifluoroethoxy benzoicacid, 2,5-bis 2,2,2-trifluorethoxy benzoic acid, 2,5-bis 2,2,2-trifluoroethoxy benzoicacid
PubChem CID 2736078
IUPAC Name 2,5-bis(2,2,2-trifluoroethoxy)benzoic acid
SMILES C1=CC(=C(C=C1OCC(F)(F)F)C(=O)O)OCC(F)(F)F

Specifications

Color White
Quantity 25 g
Formula Weight 318.17
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2,5-Bis(2,2,2-trifluoroethoxy)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 609-132-9
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.