Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Bis(2-octyldodecyl)-3,6-bis[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]pyrrolo[3,4-c]pyrrole- 98+%, TCI America™
SDP

Catalog No. B5486200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5486200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B5486200MG Supplier TCI America Supplier No. B5486200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1351986-34-9
Molecular Formula C66H110B2N2O6S2
Molecular Weight (g/mol) 1113.354
InChI Key OZQFATKGMJQJSF-UHFFFAOYSA-N
PubChem CID 86698272
IUPAC Name 2,5-bis(2-octyldodecyl)-1,4-bis[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]pyrrolo[3,4-c]pyrrole-3,6-dione
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(S2)C3=C4C(=C(N(C4=O)CC(CCCCCCCC)CCCCCCCCCC)C5=CC=C(S5)B6OC(C(O6)(C)C)(C)C)C(=O)N3CC(CCCCCCCC)CCCCCCCCCC

Specifications

Melting Point 168°C
Color Red
Quantity 200 mg
Formula Weight 1113.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Bis(2-octyldodecyl)-3,6-bis[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]pyrrolo
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.