Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Bis(4-diethylaminophenyl)-1,3,4-oxadiazole 98.0+%, TCI America™
SDP

Catalog No. B106625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B106625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B106625G Supplier TCI America Supplier No. B106625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An organic building block used in chemical research and synthesis applications.

Chemical Identifiers

CAS 1679-98-7
Molecular Formula C22H28N4O
Molecular Weight (g/mol) 364.493
MDL Number MFCD00010070
InChI Key UZGVMZRBRRYLIP-UHFFFAOYSA-N
Synonym 2,5-bis 4-diethylaminophenyl-1,3,4-oxadiazole, 2,5-bis 4'-diethylaminophenyl-1,3,4-oxadiazole, benzenamine, 4,4'-1,3,4-oxadiazole-2,5-diyl bis n,n-diethyl, 4-5-4-diethylamino phenyl-1,3,4-oxadiazol-2-yl-n,n-diethylaniline, 4-5-4-diethylamino phenyl 1,3,4-oxadiazol-2-yl phenyl diethylamine, acmc-209dxi, 2,5-bis 4-diethylaminophenyl-1,3,4-oxadiazol, 2,5-bis 4-diethylaminophenyl-1,3,4-oxadizole
PubChem CID 74301
IUPAC Name 4-[5-[4-(diethylamino)phenyl]-1,3,4-oxadiazol-2-yl]-N,N-diethylaniline
SMILES CCN(CC)C1=CC=C(C=C1)C2=NN=C(O2)C3=CC=C(C=C3)N(CC)CC

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 25 g
Formula Weight 364.49
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Bis(4-diethylaminophenyl)-1,3,4-oxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.