Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Bis(5-tert-butyl-benzoxazol-2-yl)thiophene, Sigma-Aldrich

Catalog No. 01259023 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-023 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-259-023 Supplier MilliporeSigma Supelco Supplier No. 2239995G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,5-Bis(5-tert-butyl-benzoxazol-2-yl)thiophene is a fluorescent optical brightener.

  • Suitable for photoluminescence experiments with doped silica films.
  • Used to visualize UV-patterned acrylic pressure-sensitive adhesives.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7128-64-5

Specifications

Formulation Powder
Quantity 5 g
Product Type Dye
Purity 99%
Label Type Other Labels or Dyes
Color Green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.