Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Bis(methoxycarbonyl)-3,4-diphenylcyclopentadienone, TCI America™
SDP

Catalog No. B19621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B19621G Supplier TCI America Supplier No. B19621G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16691-79-5
Molecular Formula C21H16O5
Molecular Weight (g/mol) 348.35
MDL Number MFCD00111095
InChI Key NTXFYWYVVGMAGT-UHFFFAOYSA-N
Synonym Dimethyl 3,4-Diphenylcyclopentadienone-2,5-dicarboxylate, 3,4-Diphenylcyclopentadienone-2,5-dicarboxylic Acid Dimethyl Ester
PubChem CID 2825971
IUPAC Name 1,3-dimethyl 2-oxo-4,5-diphenylcyclopenta-1(5),3-diene-1,3-dicarboxylate
SMILES COC(=O)C1=C(C(=C(C(=O)OC)C1=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 348.35
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Bis(methoxycarbonyl)-3,4-diphenylcyclopentadienone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.