Learn More
Medchemexpress LLC 2,5-bis[(morpholin-4-yl)methyl]benzene-1,4-diol | 6616-56-4 | MFCD00087356 | 99.7% | 308.37 g/mol | C16H24N2O4 | 100 MG
SDP
Catalog No. 5000246935
Encompass_Preferred
MSX-127 is a small-molecule CXCR4 antagonist used in research to study CXCR4-mediated signaling and cancer metastasis. It is a di-morpholinomethyl-substituted hydroquinone (C16H24N2O4; MW 308.37 g/mol) supplied as a high-purity solid and commonly prepared in DMSO for in vitro assays.
  • Potent CXCR4 antagonist for research applications
  • Reported to inhibit cancer metastasis in preclinical studies
  • High purity (≈99.7%) suitable for biochemical and cell-based assays
  • Provided as a powder, with solution formulations available in DMSO
  • Chemical formula C16H24N2O4; molecular weight 308.37 g/mol
Catalog No. 50-002-46935 Supplier Medchemexpress LLC Supplier No. HY103009100MG
MSX-127 is a small-molecule CXCR4 antagonist used in research to study CXCR4-mediated signaling and cancer metastasis. It is a di-morpholinomethyl-substituted hydroquinone (C16H24N2O4; MW 308.37 g/mol) supplied as a high-purity solid and commonly prepared in DMSO for in vitro assays.
  • Potent CXCR4 antagonist for research applications
  • Reported to inhibit cancer metastasis in preclinical studies
  • High purity (≈99.7%) suitable for biochemical and cell-based assays
  • Provided as a powder, with solution formulations available in DMSO
  • Chemical formula C16H24N2O4; molecular weight 308.37 g/mol