Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,5-bis(morpholin-4-ylmethyl)benzene-1,4-diol | 6616-56-4 | 99.7% | 308.37 g/mol | C16H24N2O4 | 1 ML
SDP

Catalog No. 5000246071
Encompass_Preferred

MSX-127 is a small-molecule CXCR4 antagonist supplied as a 10 mM solution in DMSO for research use. It has been reported to inhibit cancer metastasis and is provided at high purity for laboratory applications.

  • CXCR4 antagonist with reported anti-metastatic activity.
  • Supplied as a 10 mM solution in DMSO for ready-to-use dosing.
  • High purity suitable for research use (reported 99.68%).
  • Molecular weight 308.37 g/mol; chemical formula C16H24N2O4.
  • Storage guidance: in solvent, store at -80°C for long term or -20°C for short term.

Catalog No. 50-002-46071 Supplier Medchemexpress LLC Supplier No. HY10300910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MSX-127 is a small-molecule CXCR4 antagonist supplied as a 10 mM solution in DMSO for research use. It has been reported to inhibit cancer metastasis and is provided at high purity for laboratory applications.

  • CXCR4 antagonist with reported anti-metastatic activity.
  • Supplied as a 10 mM solution in DMSO for ready-to-use dosing.
  • High purity suitable for research use (reported 99.68%).
  • Molecular weight 308.37 g/mol; chemical formula C16H24N2O4.
  • Storage guidance: in solvent, store at -80°C for long term or -20°C for short term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.