Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Bis(trifluoromethyl)-3,6-dioxaundecafluorononanoyl Fluoride 95.0+%, TCI America™
SDP

Catalog No. B16985G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B16985G Supplier TCI America Supplier No. B16985G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2641-34-1
Molecular Formula C9F18O3
Molecular Weight (g/mol) 498.07
MDL Number MFCD08458100
InChI Key YSIGVPOSKQLNTO-UHFFFAOYNA-N
PubChem CID 102863
IUPAC Name 2,3,3,3-tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propanoyl fluoride
SMILES FC(=O)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
UN Number 3265
Formula Weight 498.07
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Bis(trifluoromethyl)-3,6-dioxaundecafluorononanoyl Fluoride
RTECSNumber UC3957000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.