Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Bis(trifluoromethyl)benzoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. B39805G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39805G 5 g
B39801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39805G Supplier TCI America Supplier No. B39805G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 393-82-8
Molecular Formula C9H3ClF6O
Molecular Weight (g/mol) 276.562
MDL Number MFCD00074997
InChI Key LRJNPOCQOZWIGR-UHFFFAOYSA-N
PubChem CID 2737812
IUPAC Name 2,5-bis(trifluoromethyl)benzoyl chloride
SMILES C1=CC(=C(C=C1C(F)(F)F)C(=O)Cl)C(F)(F)F

Specifications

Color Colorless
Boiling Point 72°C
Quantity 5 g
UN Number 3265
Formula Weight 276.56
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,5-Bis(trifluoromethyl)benzoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.