Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Bis[(trimethylsilyl)ethynyl]thieno[3,2-b]thiophene 98.0+%, TCI America™
SDP

Catalog No. B5196200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5196200MG 200 mg
B51961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5196200MG Supplier TCI America Supplier No. B5196200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 139896-65-4
Molecular Formula C16H20S2Si2
Molecular Weight (g/mol) 332.63
MDL Number MFCD29089350
InChI Key OWXZBWIZKNUJOL-UHFFFAOYSA-N
PubChem CID 85633934
IUPAC Name trimethyl(2-{5-[2-(trimethylsilyl)ethynyl]thieno[3,2-b]thiophen-2-yl}ethynyl)silane
SMILES C[Si](C)(C)C#CC1=CC2=C(S1)C=C(S2)C#C[Si](C)(C)C

Specifications

Melting Point 213°C
Color Yellow
Quantity 200 mg
Formula Weight 332.63
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Bis[(trimethylsilyl)ethynyl]thieno[3,2-b]thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.