Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Bis(trimethylstannyl)thieno[3,2-b]thiophene 98.0+%, TCI America™
SDP

Catalog No. B45361G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45361G 1 g
B4536200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45361G Supplier TCI America Supplier No. B45361G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 469912-82-1
Molecular Formula C12H20S2Sn2
Molecular Weight (g/mol) 465.83
MDL Number MFCD22200503
InChI Key HDZULVYGCRXVNQ-UHFFFAOYSA-N
PubChem CID 12978043
IUPAC Name trimethyl[5-(trimethylstannyl)thieno[3,2-b]thiophen-2-yl]stannane
SMILES C[Sn](C)(C)C1=CC2=C(S1)C=C(S2)[Sn](C)(C)C

Specifications

Melting Point 129°C
Quantity 1 g
UN Number 3146
Formula Weight 465.83
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Bis(trimethylstannyl)thieno[3,2-b]thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.