Learn More
AdipoGen 2 5-Bistrifluoromethylanilin
SDP
Supplier:  AdipoGen CDXB0453G025
Encompass_Preferred
Chemical. CAS 328-93-8. Formula C8H5F6N. MW 229.12. Disubstituted trifluoromethylbenzene used as a fluorinated building block or intermediate in chemical synthesis.
Catalog No. 50-256-1590