Learn More
Medchemexpress LLC 2-((5-bromo-2-((1-(methylsulfonyl)piperidin-4-yl)amino)pyrimidin-4-yl)amino)-6-fluorobenzamide | 2488892-01-7 | 99.4% | 487.35 g/mol | C17H20BrFN6O3S | 10 MG
SDP
Catalog No. 5000287302
Encompass_Preferred
TMX-2039 is a research-grade pan-cyclin-dependent kinase (pan-CDK) inhibitor intended for biochemical and cellular studies of cell cycle and transcriptional CDKs. Supplied as an off-white to light yellow solid with high reported purity, it is suitable for assay development, mechanism-of-action experiments, and probe validation in research settings.
  • Pan-CDK activity covering cell cycle and transcriptional CDKs.
  • High purity (≈99.4%) suitable for research applications.
  • Supplied as a solid powder for flexible formulation and dosing.
  • Available in small research-friendly pack sizes, e.g. 10 mg.
  • Stable under recommended storage: 4°C under nitrogen; in solvent, -80°C for 6 months or -20°C for 1 month.
  • Provides consistent material for biochemical and cellular assays.
Catalog No. 50-002-87302 Supplier Medchemexpress LLC Supplier No. HY15106910MG
TMX-2039 is a research-grade pan-cyclin-dependent kinase (pan-CDK) inhibitor intended for biochemical and cellular studies of cell cycle and transcriptional CDKs. Supplied as an off-white to light yellow solid with high reported purity, it is suitable for assay development, mechanism-of-action experiments, and probe validation in research settings.
  • Pan-CDK activity covering cell cycle and transcriptional CDKs.
  • High purity (≈99.4%) suitable for research applications.
  • Supplied as a solid powder for flexible formulation and dosing.
  • Available in small research-friendly pack sizes, e.g. 10 mg.
  • Stable under recommended storage: 4°C under nitrogen; in solvent, -80°C for 6 months or -20°C for 1 month.
  • Provides consistent material for biochemical and cellular assays.