Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(5-Bromo-2-fluorophenyl)acetonitrile, 96%, Thermo Scientific™

Catalog No. AAH62213MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62213MD 250 mg
AAH6221303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62213MD Supplier Thermo Scientific Chemicals Supplier No. H62213MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 305800-60-6
Molecular Formula C8H5BrFN
Molecular Weight (g/mol) 214.04
MDL Number MFCD09804785
InChI Key CVMIQENCTVIYOH-UHFFFAOYSA-N
Synonym 2-5-bromo-2-fluorophenyl acetonitrile, 5-bromo-2-fluorophenylacetonitrile, 5-bromo-2-fluorophenyl acetonitrile, benzeneacetonitrile, 5-bromo-2-fluoro, ksc821c8p, 5-bromo-2-fluorobenzeneacetonitrile, 5-bromo-2-fluoro-phenyl-acetonitrile
PubChem CID 23509631
IUPAC Name 2-(5-bromo-2-fluorophenyl)acetonitrile
SMILES FC1=C(CC#N)C=C(Br)C=C1

Specifications

Quantity 250 mg
UN Number UN3439
Formula Weight 214.03
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-(5-Bromo-2-fluorophenyl)acetonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.