Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol 98.0+%, TCI America™
SDP

Catalog No. B10811G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10811G 1 g
B1081100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B10811G Supplier TCI America Supplier No. B10811G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14337-53-2
Molecular Formula C15H17BrN4O
Molecular Weight (g/mol) 349.23
MDL Number MFCD00006255
InChI Key LYVSIKOGJUDRBK-QGOAFFKASA-N
Synonym 2-5-bromo-2-pyridylazo-5-diethylamino phenol, 2-5-bromopyridin-2-yl diazenyl-5-diethylamino phenol, bromo-padap, 2-5-bromo-2-pyridylazo-5-diethylaminophenol, phenol, 2-5-bromo-2-pyridinyl azo-5-diethylamino, 2-4-diethylamino-2-hydroxyphenylazo-5-bromopyridine, hnvcxdavehoibp-vhebqxmusa-n, 2-5-bromo-2-pyridylazo-5-diethylamino-phenol, 5-diethylamino-2-5-bromo 2-pyridyl diazenyl phenol, e-2-5-bromopyridin-2-yl diazenyl-5-diethylamino phenol
PubChem CID 5911417
IUPAC Name (6E)-6-[2-(5-bromopyridin-2-yl)hydrazin-1-ylidene]-3-(diethylamino)cyclohexa-2,4-dien-1-one
SMILES CCN(CC)C1=CC(=O)\C(C=C1)=N\NC1=CC=C(Br)C=N1

Specifications

Melting Point 157°C
Quantity 1 g
Formula Weight 349.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.