Learn More
2-(5-Bromo-2-pyridylazo)-5-[N-propyl-N-(3-sulfopropyl)amino]phenol disodium salt dihydrate, ≥95.0%, MilliporeSigma™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
| Linear Formula | C17H19 BrN4 Na2O4 S · 2H2 O |
| Quantity | 100 mg |
| Formula Weight | 537.34 |
| Percent Purity | ≥95.0% (CHN); ≥95.0% |
| Grade | For Spectrophotometric Det. of Zn(II), Cu(II), Fe(II), Co(II), H2O2 |
| Physical Form | Powder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.