Learn More
Medchemexpress LLC 2-(5-chloro-2-methyl-1H-indol-3-yl)-N,N-dimethylethanamine | 1210-81-7 | MFCD22201006 | 99.7% | 236.74 g/mol | C13H17ClN2 | 10 MG
SDP
Catalog No. 5000247685
Encompass_Preferred
ST1936 is a selective, nanomolar-affinity 5-HT6 receptor agonist used in pharmacological and neurobiological research. It is supplied as a solid and is available as a DMSO solution for in vitro assays. The compound has CAS 1210-81-7, molecular formula C13H17ClN2, molecular weight 236.74 g/mol, and is reported at high purity.
Catalog No. 50-002-47685 Supplier Medchemexpress LLC Supplier No. HY10311010MG
ST1936 is a selective, nanomolar-affinity 5-HT6 receptor agonist used in pharmacological and neurobiological research. It is supplied as a solid and is available as a DMSO solution for in vitro assays. The compound has CAS 1210-81-7, molecular formula C13H17ClN2, molecular weight 236.74 g/mol, and is reported at high purity.