Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(5-cyclopropyl-1-(4-ethoxy-2,6-difluorobenzyl)-4-methylpyrazol-3-yl)-5-methoxy-N-(4-pyridyl)pyrimidin-4-amine | 1445830-50-1 | 492.52 g·mol⁻¹ | C26H26F2N6O2 | 100 MG
SDP

Catalog No. 5000243642
Encompass_Preferred

BAY-320 is a selective, ATP-competitive inhibitor of the Bub1 serine/threonine kinase supplied as a solid for research use. It inhibits Bub1 with a reported IC50 of about 680 nM and is used to study mitotic chromosome arrangement, spindle assembly, and cancer cell proliferation. The compound is provided as a powder with recommended low-temperature storage for long-term stability.

  • Selective ATP-competitive Bub1 kinase inhibitor.
  • Reported IC50 ~680 nM against Bub1.
  • Solid powder suitable for biochemical and cellular assays.
  • Molecular weight 492.52 g·mol⁻¹; formula C26H26F2N6O2.
  • Recommended storage: powder -20 °C (3 years), 4 °C (2 years).
  • Useful for studies of mitosis and cancer cell proliferation.

Catalog No. 50-002-43642 Supplier Medchemexpress LLC Supplier No. HY104000100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BAY-320 is a selective, ATP-competitive inhibitor of the Bub1 serine/threonine kinase supplied as a solid for research use. It inhibits Bub1 with a reported IC50 of about 680 nM and is used to study mitotic chromosome arrangement, spindle assembly, and cancer cell proliferation. The compound is provided as a powder with recommended low-temperature storage for long-term stability.

  • Selective ATP-competitive Bub1 kinase inhibitor.
  • Reported IC50 ~680 nM against Bub1.
  • Solid powder suitable for biochemical and cellular assays.
  • Molecular weight 492.52 g·mol⁻¹; formula C26H26F2N6O2.
  • Recommended storage: powder -20 °C (3 years), 4 °C (2 years).
  • Useful for studies of mitosis and cancer cell proliferation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.