Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Di(2-thienyl)-1H-pyrrole 95.0+%, TCI America™
SDP

Catalog No. D38761G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D38761G Supplier TCI America Supplier No. D38761G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 89814-62-0
Molecular Formula C12H9NS2
Molecular Weight (g/mol) 231.331
InChI Key REHRCHHNCOTPBV-UHFFFAOYSA-N
PubChem CID 4175110
IUPAC Name 2,5-dithiophen-2-yl-1H-pyrrole
SMILES C1=CSC(=C1)C2=CC=C(N2)C3=CC=CS3

Specifications

Melting Point 84°C
Color Yellow
Quantity 1 g
Formula Weight 231.33
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Di(2-thienyl)-1H-pyrrole
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.