Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,5-Diaminobenzenesulfonic acid

Catalog No. 3277650G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-776-50G 50 g
32-776-250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 32-776-50G Supplier Sigma Aldrich Supplier No. 3277650G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,5-Diaminobenzenesulfonic acid

Specifications

CAS 88-45-9
Melting Point 298°C to 300°C (dec.) (lit.)
Molecular Formula C6H8N2O3S
Linear Formula (H2N)2C6H3SO3H
MDL Number MFCD00007904
Quantity 50 g
Molecular Weight (g/mol) 188.2
Percent Purity ≥97.0% (T)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.