Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibenzylidenecyclopentanone 98.0+%, TCI America™
SDP

Catalog No. D35975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35975G 5 g
D359725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35975G Supplier TCI America Supplier No. D35975G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 895-80-7
Molecular Formula C19H16O
Molecular Weight (g/mol) 260.336
MDL Number MFCD00019321
InChI Key CVTOCKKPVXJIJK-SIJTWYJSSA-N
PubChem CID 6436781
IUPAC Name (2Z,5E)-2,5-dibenzylidenecyclopentan-1-one
SMILES C1CC(=CC2=CC=CC=C2)C(=O)C1=CC3=CC=CC=C3

Specifications

Melting Point 192°C
Color Yellow
Quantity 5 g
Formula Weight 260.34
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibenzylidenecyclopentanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.