Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromo-1,1,3,4-tetraphenylsilole 98.0+%, TCI America™
SDP

Catalog No. D43101G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43101G 1 g
D43105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43101G Supplier TCI America Supplier No. D43101G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 200068-36-6
Molecular Formula C28H20Br2Si
Molecular Weight (g/mol) 544.36
MDL Number MFCD28100827
InChI Key MXGIPLZPCLLOTQ-UHFFFAOYSA-N
Synonym 2,5-Dibromo-1,1,3,4-tetraphenylsilacyclopenta-2,4-diene
PubChem CID 15321324
IUPAC Name 2,5-dibromo-1,1,3,4-tetraphenyl-1H-silole
SMILES BrC1=C(C(=C(Br)[Si]1(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 170°C
Quantity 1 g
Formula Weight 544.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromo-1,1,3,4-tetraphenylsilole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.