Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromo-3-hexadecylthiophene 96.0+%, TCI America™
SDP

Catalog No. D52425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52425G 5 g
D52421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52425G Supplier TCI America Supplier No. D52425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 178452-13-6
Molecular Formula C20H34Br2S
Molecular Weight (g/mol) 466.36
MDL Number MFCD30742899
InChI Key KBLOMHRWQCLPTC-UHFFFAOYSA-N
PubChem CID 101022660
IUPAC Name 2,5-dibromo-3-hexadecylthiophene
SMILES CCCCCCCCCCCCCCCCC1=C(Br)SC(Br)=C1

Specifications

Melting Point 36°C
Color White-Yellow
Quantity 5 g
Formula Weight 466.36
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromo-3-hexadecylthiophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.