Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,5-Dibromo-3-hexylthiophene

Catalog No. 4563735G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-637-35G 5 g
45-637-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-637-35G Supplier Sigma Aldrich Supplier No. 4563735G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,5-Dibromo-3-hexylthiophene is a 2,5 coupled conductive polymer with conjugated polythiophene based system, which has a controllable band gap.It can be used as a monomer with 5,5G-dibromo-3,3G-dihexyl-2,2G-bithiophene, which can synthesize regioregular-P3HT-regiosymmetric-P3HT (a diblock polymer) for organic electronics based applications. It can also be used in the synthesis of P3HT, which can be potentially used for photocatalytic applications.Product of Rieke Metals, Inc. Rieke is a registered trademark of Rieke Metals, Inc.

Specifications

CAS 116971-11-0
Density 1.521 g/mL (at 25°C)
Molecular Formula C10H14Br2S
Refractive Index n20/D 1.557 (literature)
Linear Formula C10H14Br2S
Quantity 5 g
MDL Number MFCD00274313
Synonym 2,5-Dibromo-3-hex-1-ylthiophene
Molecular Weight (g/mol) 326.09
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.