Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromo-3-hexylthiophene 97.0+%, TCI America™
SDP

Catalog No. D38965G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. D38965G Supplier TCI America Supplier No. D38965G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 116971-11-0
Molecular Formula C10H14Br2S
Molecular Weight (g/mol) 326.09
MDL Number MFCD00274313
InChI Key NSYFIAVPXHGRSH-UHFFFAOYSA-N
PubChem CID 3936632
IUPAC Name 2,5-dibromo-3-hexylthiophene
SMILES CCCCCCC1=C(Br)SC(Br)=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 326.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dibromo-3-hexylthiophene
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.