Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromo-4-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. D40585G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40585G 5 g
D40581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40585G Supplier TCI America Supplier No. D40585G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3430-26-0 | C6H5Br2N | 250.921 g/mol

Chemical Identifiers

CAS 3430-26-0
Molecular Formula C6H5Br2N
Molecular Weight (g/mol) 250.921
MDL Number MFCD00234955
InChI Key WWJLJUAHQHXDGM-UHFFFAOYSA-N
PubChem CID 2734429
IUPAC Name 2,5-dibromo-4-methylpyridine
SMILES CC1=CC(=NC=C1Br)Br

Specifications

Melting Point 40°C
Color Yellow
Quantity 5 g
Synonym 2,5-dibromo-4-picoline, pyridine, 2,5-dibromo-4-methyl, 2,5-dibromo-4-methyl-pyridine, pubchem2411, 2 pound not5-dibromo-4-methylpyridine, acmc-209i6i, 2, 5-dibromo-4-picoline, 2,5-dibromo-4-methypyridine, ksc495g9l, tpc-py104
Formula Weight 250.92
Percent Purity ≥98.0% (GC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromo-4-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.