Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromobenzenesulfonyl chloride, 99%

Catalog No. AAL1988806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1988806 5 g
AAL1988803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1988806 Supplier Thermo Scientific Chemicals Supplier No. L1988806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 23886-64-8 | C6H3Br2ClO2S | 334.406 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23886-64-8
Molecular Formula C6H3Br2ClO2S
Molecular Weight (g/mol) 334.406
MDL Number MFCD00060685
InChI Key ZLMPLIWURYRGEB-UHFFFAOYSA-N
Synonym 2,5-dibromobenzene-1-sulfonyl chloride, 2,5-dibromobenzenesulphonyl chloride, benzenesulfonyl chloride, 2,5-dibromo, 2,5-dibromophenyl chlorosulfone, 2,5-dibromobenzene sulphonyl chloride, pubchem5112, acmc-1cbvd, ksc201m8p, 2,5-dibromophenylsulfonyl chloride, 2,5-dibromobenzenesulphonylchloride
PubChem CID 2737515
IUPAC Name 2,5-dibromobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1Br)S(=O)(=O)Cl)Br

Specifications

Melting Point 69°C to 72°C
Quantity 5 g
UN Number UN3261
Beilstein 2694054
Sensitivity Moisture sensitive
Formula Weight 334.41
Percent Purity 99%
Chemical Name or Material 2,5-Dibromobenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.