Learn More
Medchemexpress LLC 2,5-dibromohexanediamide | 99584-96-0 | MFCD27976335 | ≥95.0% | 301.96 g/mol | C6H10Br2N2O2 | 250 MG
SDP
Catalog No. 5000248136
Encompass_Preferred
DBHDA (2,5-dibromohexanediamide) is a small organic reagent used to convert cysteine residues to dehydroalanine (DHA) in peptides and proteins. Supplied as a solid powder for research applications, it supports selective chemical modification workflows in protein engineering and bioconjugation studies. Follow supplier guidance for storage and handling.
Catalog No. 50-002-48136 Supplier Medchemexpress LLC Supplier No. HY101847250MG
DBHDA (2,5-dibromohexanediamide) is a small organic reagent used to convert cysteine residues to dehydroalanine (DHA) in peptides and proteins. Supplied as a solid powder for research applications, it supports selective chemical modification workflows in protein engineering and bioconjugation studies. Follow supplier guidance for storage and handling.