Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,5-Dibromohexanediamide

Catalog No. 900607250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-060-7250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 90-060-7250MG Supplier Sigma Aldrich Supplier No. 900607250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This compound has been shown to be a useful reagent for the conversion of cysteine to dehydroalanine (DHA) in peptides or proteins. This enables chemical mutagenesis in which DHA can be efficiently reacted with iodide building blocks to add various natural and unnatural side chains on proteins. It was also shown that other modifications could be added through DHA such as phosphorylation, methylation, and glycosylation. In other research it was shown that DHA can be used to make ubiquitin conjugates that have a bond that mimics the native isopeptide bond.

Specifications

CAS 42969-65-3
Density 0.89 g/mL at 25°C
Boiling Point 108°C to 111°C
Molecular Formula C4H6O
Linear Formula HC///CCH(OH)CH3
Quantity 250 mg
MDL Number MFCD00211237
Molecular Weight (g/mol) 70.09 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.