Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromopyrimidine 98.0+%, TCI America™
SDP

Catalog No. D42255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42255G 5 g
D42251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42255G Supplier TCI America Supplier No. D42255G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32779-37-6
Molecular Formula C4H2Br2N2
Molecular Weight (g/mol) 237.88
MDL Number MFCD08275684
InChI Key XAHITOJPIWZJHD-UHFFFAOYSA-N
PubChem CID 10955588
IUPAC Name 2,5-dibromopyrimidine
SMILES BrC1=CN=C(Br)N=C1

Specifications

Melting Point 85°C
Color White-Yellow
Quantity 5 g
Formula Weight 237.88
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromopyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.