Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromothieno[3,2-b]thiophene 98.0+%, TCI America™
SDP

Catalog No. D36781G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36781G 1 g
D3678200MG 200 mg
D36785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D36781G Supplier TCI America Supplier No. D36781G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25121-87-3
Molecular Formula C6H2Br2S2
Molecular Weight (g/mol) 298.01
MDL Number MFCD03931293
InChI Key APDAUBNBDJUQGW-UHFFFAOYSA-N
PubChem CID 3597455
IUPAC Name 2,5-dibromothieno[3,2-b]thiophene
SMILES C1=C(SC2=C1SC(=C2)Br)Br

Specifications

Melting Point 130°C
Quantity 1 g
Formula Weight 298.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromothieno[3,2-b]thiophene
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.