Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromothiophene 1,1-Dioxide 98.0+%, TCI America™
SDP

Catalog No. D43761G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D43761G Supplier TCI America Supplier No. D43761G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 89088-95-9
Molecular Formula C4H2Br2O2S
Molecular Weight (g/mol) 273.926
InChI Key LKJJLFANVUXGGB-UHFFFAOYSA-N
PubChem CID 10923739
IUPAC Name 2,5-dibromothiophene 1,1-dioxide
SMILES C1=C(S(=O)(=O)C(=C1)Br)Br

Specifications

Melting Point 126°C
Color White-Yellow
Quantity 1 g
Formula Weight 273.93
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromothiophene 1,1-Dioxide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.