Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromothiophene, MP Biomedicals

Catalog No. ICN212977 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN212977 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN212977 Supplier MP Biomedicals, Inc Supplier No. 0521297725

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3141-27-3
Molecular Formula C4H2Br2S
Molecular Weight (g/mol) 241.93
MDL Number MFCD00005420
InChI Key KBVDUUXRXJTAJC-UHFFFAOYSA-N
Synonym thiophene, 2,5-dibromo, 2,5-dibromo-thiophene, unii-f0033k8cxm, pubchem5512, thiophene,5-dibromo, acmc-1ckmh, 2,5-bis bromanyl thiophene, ksc224g0j, 2,5-dibromothiophene, bidd:gt0086
PubChem CID 18453
IUPAC Name 2,5-dibromothiophene
SMILES BrC1=CC=C(Br)S1

Specifications

Quantity 25 g
Formula Weight 241.94
Chemical Name or Material 2, 5-Dibromothiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.