Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichloro-3-nitropyridine, ≥97%, Thermo Scientific™

Catalog No. AAH6427622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6427622 100 g
AAH6427614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6427622 Supplier Thermo Scientific Chemicals Supplier No. H6427622

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21427-62-3
Molecular Formula C5H2Cl2N2O2
Molecular Weight (g/mol) 192.983
MDL Number MFCD06658963
InChI Key OBUGJYJQJWMOQO-UHFFFAOYSA-N
PubChem CID 8063331
IUPAC Name 2,5-dichloro-3-nitropyridine
SMILES C1=C(C(=NC=C1Cl)Cl)[N+](=O)[O-]

Specifications

Melting Point 41°C to 45°C
Flash Point >110°C (230°F)
Quantity 100 g
UN Number UN2811
Beilstein 147539
Synonym 2,5-dichloro-3-nitro-pyridine, pyridine, 2,5-dichloro-3-nitro, 2.5-dichloro-3-nitropyridine, 2,5-dichloro-3-nitro pyridine, pubchem2267, acmc-1cdlk, 2,5dichloro-3-nitropyridine, ksc495e7t, 2,5 dichloro-3-nitropyridine, 2,5 dichloro-3-nitro pyridine
Formula Weight 192.98
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material 2,5-Dichloro-3-nitropyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.