Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichloro-3-nitropyridine 98.0+%, TCI America™
SDP

Catalog No. D42675G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42675G 5 g
D426725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42675G Supplier TCI America Supplier No. D42675G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 21427-62-3
Molecular Formula C5H2Cl2N2O2
Molecular Weight (g/mol) 192.983
MDL Number MFCD06658963
InChI Key OBUGJYJQJWMOQO-UHFFFAOYSA-N
PubChem CID 8063331
IUPAC Name 2,5-dichloro-3-nitropyridine
SMILES C1=C(C(=NC=C1Cl)Cl)[N+](=O)[O-]

Specifications

Melting Point 45°C
Quantity 5 g
Synonym 2,5-dichloro-3-nitro-pyridine, pyridine, 2,5-dichloro-3-nitro, 2.5-dichloro-3-nitropyridine, 2,5-dichloro-3-nitro pyridine, pubchem2267, acmc-1cdlk, 2,5dichloro-3-nitropyridine, ksc495e7t, 2,5 dichloro-3-nitropyridine, 2,5 dichloro-3-nitro pyridine
Formula Weight 192.98
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichloro-3-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.