Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',5'-Dichloroacetanilide 98.0+%, TCI America™
SDP

Catalog No. D030725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D030725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D030725G Supplier TCI America Supplier No. D030725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2621-62-7
Molecular Formula C8H7Cl2NO
Molecular Weight (g/mol) 204.05
MDL Number MFCD00018519
InChI Key ICZFWTSENFTULW-UHFFFAOYSA-N
PubChem CID 17494
IUPAC Name N-(2,5-dichlorophenyl)acetamide
SMILES CC(=O)NC1=C(C=CC(=C1)Cl)Cl

Specifications

Melting Point 135°C
Color White
Quantity 25 g
Formula Weight 204.05
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2′,5′-Dichloroacetanilide
EINECSNumber (3)-0262
RTECSNumber AE1965000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.