Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',5'-Dichloroacetophenone, 98%

Catalog No. AAA1377009
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1377009 10 g
AAA1377014 25 g
AAA1377022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1377009 Supplier Thermo Scientific Chemicals Supplier No. A1377009
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 2476-37-1 | C8H6Cl2O | 189.035 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2476-37-1
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.035
MDL Number MFCD00000607
InChI Key CYNFEPKQDJHIMV-UHFFFAOYSA-N
Synonym 2',5'-dichloroacetophenone, 1-2,5-dichlorophenyl ethanone, 2,5-dichloroacetophenone, ethanone, 1-2,5-dichlorophenyl, acetophenone, 2',5'-dichloro, 1-2,5-dichlorophenyl ethan-1-one, 1-acetyl-2,5-dichlorobenzene, 2,5-dichloracetophenon, pubchem10586, acmc-1cmfm
PubChem CID 75587
IUPAC Name 1-(2,5-dichlorophenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1)Cl)Cl

Specifications

Melting Point 11°C to 13°C
Density 1.312
Boiling Point 118°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.561
Quantity 10 g
Beilstein 1865043
Formula Weight 189.04
Percent Purity 98%
Chemical Name or Material 2',5'-Dichloroacetophenone
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 219-605-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.