Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichlorobenzaldehyde, 98%

Catalog No. AAA1352514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1352514 25 g
AAA1352506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1352514 Supplier Thermo Scientific Chemicals Supplier No. A1352514
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6361-23-5 | C7H4Cl2O | 175.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6361-23-5
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.01
MDL Number MFCD00156140
InChI Key BUXHYMZMVMNDMG-UHFFFAOYSA-N
Synonym 2,5-dichloro-benzaldehyde, 2,5-dichlorobenzaldehdye, benzaldehyde, 2,5-dichloro, pubchem3081, 2.5-dichlorbenzaldehyd, 2,5-dichlorbenzaldehyde, acmc-1bkcw, 2,5-dichloro benzaldehdye, ksc493o7f, 2,5-dichlorobenzaldehyde
PubChem CID 80702
IUPAC Name 2,5-dichlorobenzaldehyde
SMILES ClC1=CC=C(Cl)C(C=O)=C1

Specifications

Melting Point 53°C to 58°C
Boiling Point 231°C to 233°C
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN1759
Beilstein 1635476
Sensitivity Air Sensitive
Formula Weight 175.01
Percent Purity 98%
Chemical Name or Material 2,5-Dichlorobenzaldehyde
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber 228-832-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.