Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichlorobenzylmagnesium chloride, 0.25M in 2-MeTHF, Thermo Scientific™

Catalog No. AAH54707AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54707AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH54707AE Supplier Thermo Scientific Chemicals Supplier No. H54707AE

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 213627-19-1
Molecular Formula C7H5Cl3Mg
Molecular Weight (g/mol) 219.77
MDL Number MFCD06200641
InChI Key LCSTXMZCRKYVRE-UHFFFAOYSA-M
Synonym 2,5-dichlorobenzylmagnesium chloride, 2,5-dichlorobenzylmagnesium chloride, 0.25m in 2-methf, chloro 2,5-dichlorophenyl methyl magnesium, gcykxkkuuljojj-uhfffaoysa-m, 2,5-dichlorobenzyl magnesium chloride, magnesium,1,4-dichloro-2-methanidylbenzene,chloride, 2,5-dichlorobenzylmagnesium chloride 0.25 m in diethyl ether
PubChem CID 24722686
IUPAC Name magnesium;1,4-dichloro-2-methanidylbenzene;chloride
SMILES [Mg++].[Cl-].[CH2-]C1=C(Cl)C=CC(Cl)=C1

Specifications

Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Formula Weight 219.78
Concentration or Composition (by Analyte or Components) 0.25M in 2-MeTHF
Chemical Name or Material 2,5-Dichlorobenzylmagnesium chloride
Hazard Category H225-H302-H314-H335
Hazard Statement GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.