Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichlorophenol, 98%

Catalog No. AAB2478722
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2478722 100 g
AAB2478714 25 g
AAB2478736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AAB2478722 Supplier Thermo Scientific Chemicals Supplier No. B2478722
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 583-78-8 | C6H4Cl2O | 163.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-78-8
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163.00
MDL Number MFCD00002174
InChI Key RANCECPPZPIPNO-UHFFFAOYSA-N
Synonym phenol, 2,5-dichloro, 2,5-dichloro-phenol, 2,5-dcp, unii-3b11g9akba, 3,6-dichlorophenol, ccris 5903, 3b11g9akba, dsstox_cid_5003, dsstox_rid_77621, dsstox_gsid_25003
PubChem CID 66
ChEBI CHEBI:27929
IUPAC Name 2,5-dichlorophenol
SMILES OC1=CC(Cl)=CC=C1Cl

Specifications

Melting Point 55°C to 58°C
Boiling Point 211°C
Odor Phenol-like
Quantity 100 g
UN Number UN2020
Beilstein 1907692
Formula Weight 163
Percent Purity 98%
Chemical Name or Material 2,5-Dichlorophenol
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, SOLID
EINECSNumber 209-520-4
RTECSNumber SK8600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.